6-Methoxyquinolin-8-amine 98%
Lieferant: Apollo Scientific
Synonyme:
8-Amino-6-methoxyquinoline
OR911787-5G
APOSOR911787-5GEA
55.1
EUR
APOSOR911787-5G
6-Methoxyquinolin-8-amine 98%
6-Methoxyquinolin-8-amine
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672902 CAS-Nummer: 90-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...