2,3-Dimethylanisol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methoxy-2,3-dimethylbenzene

OR911822-100G OR911822-25G
APOSOR911822-100GEA 180 EUR
APOSOR911822-100G APOSOR911822-25G
2,3-Dimethylanisol 95%
2,3-Dimethylanisol
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008376
CAS-Nummer: 2944-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR