Fmoc-L-Tyr(Me)-OH 97%

Lieferant: Apollo Scientific

Synonyme: Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine

OR911865-25G
APOSOR911865-25GEA 140 EUR
APOSOR911865-25G
Fmoc-L-Tyr(Me)-OH 97%
Fmoc-L-Tyr(Me)-OH
Formel: C₂₅H₂₃NO₅
Molecular Weight: 417,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153368
CAS-Nummer: 77128-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR