(R)-(-)-1-Indanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-(-)-1-Hydroxyindan
OR911887-1G
OR911887-250MG
APOSOR911887-1GEA
163
EUR
APOSOR911887-1G
APOSOR911887-250MG
(R)-(-)-1-Indanol 95%
(R)-(-)-1-Indanol
Formel:
C₉H₁₀O Molecular Weight: 134,18 g/mol Schmelzpunkt: 72…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064164 CAS-Nummer: 697-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...