1-(3,4-Dichlorophenyl)-N-methylmethanamine 98%

Lieferant: Apollo Scientific

Synonyme: (3,4-Dichlorobenzyl)methylamine

OR911950-25G
APOSOR911950-25GEA 171 EUR
APOSOR911950-25G
1-(3,4-Dichlorophenyl)-N-methylmethanamine 98%
1-(3,4-Dichlorophenyl)-N-methylmethanamine
Formel: C₈H₉Cl₂N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 5635-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR