3,4-Dimethoxybenzylalkohol 98%

Lieferant: Apollo Scientific

Synonyme: Veratrylalkohol, (3,4-Dimethoxyphenyl)methanol

OR911996-100G OR911996-500G
APOSOR911996-100GEA 56.7 EUR
APOSOR911996-100G APOSOR911996-500G
3,4-Dimethoxybenzylalkohol 98%
3,4-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00004638
CAS-Nummer: 93-03-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR