3-Ethoxyphenol 95%

Lieferant: Apollo Scientific

OR912018-100G OR912018-25G OR912018-5G
APOSOR912018-100GEA 519 EUR
APOSOR912018-100G APOSOR912018-25G APOSOR912018-5G
3-Ethoxyphenol 95%
3-Ethoxyphenol
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016450
CAS-Nummer: 621-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR