5,6,7,8-Tetrahydro-1-naphthalincarbonsäure 98%

Lieferant: Apollo Scientific

OR912055-25G
APOSOR912055-25GEA 126 EUR
APOSOR912055-25G
5,6,7,8-Tetrahydro-1-naphthalincarbonsäure 98%
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,2118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01464030
CAS-Nummer: 4242-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR