2-Methyl-1,2,3,4-tetrahydrochinolin 95%
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline
OR912093-100G
OR912093-25G
APOSOR912093-100GEA
307
EUR
APOSOR912093-100G
APOSOR912093-25G
2-Methyl-1,2,3,4-tetrahydrochinolin 95%
1,2,3,4-Tetrahydroquinaldine
Formel:
C₁₀H₁₃N Molecular Weight: 147,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023886 CAS-Nummer: 1780-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...