3-Nitrophenetole 95%
Lieferant: Apollo Scientific
Synonyme:
1-Ethoxy-3-nitrobenzene
OR912100-25G
APOSOR912100-25GEA
119
EUR
APOSOR912100-25G
3-Nitrophenetole 95%
3-Nitrophenetole
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024472 CAS-Nummer: 621-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...