1,4-Bis(methoxymethyl)benzol 95%

Lieferant: Apollo Scientific

Synonyme: p-Xylene Glycol Dimethyl Ether

OR912103-100G OR912103-500G
APOSOR912103-100GEA 63.8 EUR
APOSOR912103-100G APOSOR912103-500G
1,4-Bis(methoxymethyl)benzol 95%
1,4-Bis(methoxymethyl)benzene
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025903
CAS-Nummer: 6770-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR