5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanon 96%
Lieferant: Apollo Scientific
Synonyme:
5-Ethyl-3-hydroxy-4-methylfuran-2(5H)-one, Ethyl Fenugreek Lactone
OR912126-25G
APOSOR912126-25GEA
85.7
EUR
APOSOR912126-25G
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanon 96%
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanon
Formel:
C₇H₁₀O₃ Molecular Weight: 142,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036673 CAS-Nummer: 698-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...