3,5-Dimethoxybenzylamin 97%

Lieferant: Apollo Scientific

Synonyme: 3,5-Dimethoxybenzyl amine

OR912260-25G
APOSOR912260-25GEA 111 EUR
APOSOR912260-25G
3,5-Dimethoxybenzylamin 97%
3,5-Dimethoxybenzylamin
Formel: C₉H₁₃NO₂
Molecular Weight: 167,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052813
CAS-Nummer: 34967-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR