5-Amino-1,3-dihydro-2H-benzimidazol-2-on 98%
Lieferant: Apollo Scientific
Synonyme:
5-Amino-2-benzimidazolinone
OR912262-500G
APOSOR912262-500GEA
69.6
EUR
APOSOR912262-500G
5-Amino-1,3-dihydro-2H-benzimidazol-2-on 98%
5-Amino-1,3-dihydro-2H-benzimidazol-2-one
Formel:
C₇H₇N₃O Molecular Weight: 149,1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053555 CAS-Nummer: 95-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...