(S)-(+)-Methylmandelat 98%
Lieferant: Apollo Scientific
Synonyme:
(S)-(+)-2-Hydroxy-2-phenylethansäuremethylester, L(+)-Methylglykolat, L(+)-2-Hydroxy-2-phenylessigsäuremethylester, Methyl-L(+)-glykolat, (S)-(+)-Methylglykolat, (S)-(+)-2-Hydroxy-2-phenylessigsäuremethylester, L-2-Hydroxy-2-phenylethansäuremethylester, L(+)-Mandelsäuremethylester, L(+)-Methylmandelat, Methyl-(S)-(+)-glykolat, (S)-(+)-Mandelsäuremethylester
OR912288-100G
APOSOR912288-100GEA
112
EUR
APOSOR912288-100G
(S)-(+)-Methylmandelat 98%
(S)-(+)-Methylmandelat
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064246 CAS-Nummer: 21210-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...