3,5-Dimethylbenzaldehyd 98%

Lieferant: Apollo Scientific

OR912342-100G
APOSOR912342-100GEA 203 EUR
APOSOR912342-100G
3,5-Dimethylbenzaldehyd 98%
3,5-Dimethylbenzaldehyde
Formel: C₉H₁₀O
Molecular Weight: 134,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082777
CAS-Nummer: 5779-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR