(R)-(+)-α-Ethylbenzylamin 95%

Lieferant: Apollo Scientific

Synonyme: (R)-1-Phenylpropan-1-amine, (R)-(+)-1-Phenylpropylamine

OR912344-10G OR912344-25G OR912344-5G
APOSOR912344-10GEA 261 EUR
APOSOR912344-10G APOSOR912344-25G APOSOR912344-5G
(R)-(+)-α-Ethylbenzylamin 95%
(R)-(+)-α-Ethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00083057
CAS-Nummer: 3082-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR