2,3,5-Trimethylanisol 95+%
Lieferant: Apollo Scientific
Synonyme:
1-Methoxy-2,3,5-trimethylbenzene
OR912380-100G
OR912380-25G
APOSOR912380-100GEA
325
EUR
APOSOR912380-100G
APOSOR912380-25G
2,3,5-Trimethylanisol 95+%
2,3,5-Trimethylanisol
Formel:
C₁₀H₁₄O Molecular Weight: 150,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142783 CAS-Nummer: 20469-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...