6-Methoxysalicylaldehyd 95%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxy-6-methoxybenzaldehyde
OR912396-5G
APOSOR912396-5GEA
59.6
EUR
APOSOR912396-5G
6-Methoxysalicylaldehyd 95%
6-Methoxysalicylaldehyd
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151830 CAS-Nummer: 700-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...