2-Methyl-5,6,7,8-tetrahydrochinolin 95%

Lieferant: Apollo Scientific

Synonyme: 5,6,7,8-Tetrahydroquinaldine

OR912429-10G OR912429-1G OR912429-5G
APOSOR912429-10GEA 429 EUR
APOSOR912429-10G APOSOR912429-1G APOSOR912429-5G
2-Methyl-5,6,7,8-tetrahydrochinolin 95%
2-Methyl-5,6,7,8-tetrahydrochinolin
Formel: C₁₀H₁₃N
Molecular Weight: 147,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191581
CAS-Nummer: 2617-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR