3-(2-Furyl)acrolein 98%
Lieferant: Apollo Scientific
Synonyme:
3-(Furan-2-yl)acrylaldehyde, 3-(2-Furyl)propenal
OR912456-100G
APOSOR912456-100GEA
94.4
EUR
APOSOR912456-100G
3-(2-Furyl)acrolein 98%
3-(2-Furyl)acrolein
Formel:
C₇H₆O₂ Molecular Weight: 122.12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003256 CAS-Nummer: 623-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...