2-(3-Oxo-1,2,3,4-tetrahydroquinoxalin-2-yl)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 1,2,3,4-Tetrahydro-3-oxo-2-quinoxalineacetic acid

OR912458-1G OR912458-250MG OR912458-5G
APOSOR912458-1GEA 213 EUR
APOSOR912458-1G APOSOR912458-250MG APOSOR912458-5G
2-(3-Oxo-1,2,3,4-tetrahydroquinoxalin-2-yl)acetic acid 95%
2-(3-Oxo-1,2,3,4-tetrahydroquinoxalin-2-yl)acetic acid
Formel: C₁₀H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 136584-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR