Methyl-3-hydroxy-2-pyridincarboxylat 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-Hydroxy-2-pyridinecarboxylate, 3-Hydroxypyridine-2-carboxylic acid methyl ester, 3-Hydroxypicolinic acid methyll ester, Methyl 3-hydroxypicolinate, 3-Hydroxy-2-pyridinecarboxylic acid methyll ester, Methyl 3-Hydroxypicolinate
OR912546-1G
OR912546-25G
OR912546-5G
APOSOR912546-1GEA
63.8
EUR
APOSOR912546-1G
APOSOR912546-25G
APOSOR912546-5G
Methyl-3-hydroxy-2-pyridincarboxylat 98%
Methyl-3-hydroxypicolinate
Formel:
C₇H₇NO₃ Molecular Weight: 153,1354 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661311 CAS-Nummer: 62733-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...