Amino(3-hydroxyphenyl)acetic acid hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
2-(3-Hydroxyphenyl)glycine
OR912561-1G
OR912561-500MG
OR912561-5G
APOSOR912561-1GEA
348
EUR
APOSOR912561-1G
APOSOR912561-500MG
APOSOR912561-5G
Amino(3-hydroxyphenyl)acetic acid hydrochloride 97%
Amino(3-hydroxyphenyl)acetic acid hydrochloride
Formel:
C₈H₁₀ClNO₃ Molecular Weight: 203,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19103264 CAS-Nummer: 31932-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...