Methyl-2-(2-formylphenoxy)acetate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(2-formylphenoxy)acetate

OR912771-5G
APOSOR912771-5GEA 134 EUR
APOSOR912771-5G
Methyl-2-(2-formylphenoxy)acetate 95%
Methyl-2-(2-formylphenoxy)acetate
Formel: C₁₀H₁₀O₄
Molecular Weight: 194.1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02629604
CAS-Nummer: 40359-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR