4-Ethyl-6-hydroxy-2-methylpyrimidine 95%
Lieferant: Apollo Scientific
OR912842-1G
OR912842-250MG
OR912842-5G
APOSOR912842-1GEA
0
EUR
APOSOR912842-1G
APOSOR912842-250MG
APOSOR912842-5G
4-Ethyl-6-hydroxy-2-methylpyrimidine 95%
4-Ethyl-6-hydroxy-2-methylpyrimidine
Formel:
C₇H₁₀N₂O Molecular Weight: 138,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03265397 CAS-Nummer: 52421-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...