1-(3-Amino-2-chloropyridin-4-yl)ethanone 95%

Lieferant: Apollo Scientific

OR912852-1G OR912852-250MG
APOSOR912852-1GEA 432 EUR
APOSOR912852-1G APOSOR912852-250MG
1-(3-Amino-2-chloropyridin-4-yl)ethanone 95%
1-(3-Amino-2-chloropyridin-4-yl)ethanone
Formel: C₇H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03411638
CAS-Nummer: 342899-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR