4-Cyan-4'-methylbiphenyl 98%
Lieferant: Apollo Scientific
Synonyme:
4-Methyl-[1,1-biphenyl]-4-carbonitrile
OR912900-25G
OR912900-5G
APOSOR912900-25GEA
299
EUR
APOSOR912900-25G
APOSOR912900-5G
4-Cyan-4'-methylbiphenyl 98%
4-Cyan-4'-methylbiphenyl
Formel:
C₁₄H₁₁N Molecular Weight: 193,2438 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03703444 CAS-Nummer: 50670-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...