5-Chloropyrimidine 96%
Lieferant: Apollo Scientific
OR912930-10G
OR912930-1G
OR912930-5G
APOSOR912930-10GEA
388
EUR
APOSOR912930-10G
APOSOR912930-1G
APOSOR912930-5G
5-Chloropyrimidine 96%
5-Chloropyrimidine
Formel:
C4H3ClN2 Molecular Weight: 114,5300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840717 CAS-Nummer: 17180-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...