2-Chlor-3-hydroxy-4-picolin 95%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-4-methylpyridin-3-ol
OR912990-1G
OR912990-250MG
APOSOR912990-1GEA
89.9
EUR
APOSOR912990-1G
APOSOR912990-250MG
2-Chlor-3-hydroxy-4-picolin 95%
2-Chlor-3-hydroxy-4-picolin
Formel:
C₆H₆ClNO Molecular Weight: 143,5700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112564 CAS-Nummer: 884494-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...