5-(p-Tolyl)pyrimidin-2-amine 95%
Lieferant: Apollo Scientific
OR913113-1G
OR913113-250MG
APOSOR913113-1GEA
290
EUR
APOSOR913113-1G
APOSOR913113-250MG
5-(p-Tolyl)pyrimidin-2-amine 95%
5-(p-Tolyl)pyrimidin-2-amine
Formel:
C₁₁H₁₁N₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
31408-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...