3-Cyanophenylacetic acid 98%
Lieferant: Apollo Scientific
Synonyme:
m-Cyanophenylacetic acid, 2-(3-cyanophenyl)acetic Acid, Benzeneacetic acid,3-cyano-
OR913147-10G
OR913147-1G
OR913147-5G
APOSOR913147-10GEA
287
EUR
APOSOR913147-10G
APOSOR913147-1G
APOSOR913147-5G
3-Cyanophenylacetic acid 98%
3-Cyanophenylacetic acid
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411079 CAS-Nummer: 1878-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...