5-Ethoxy-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

OR913219-1G
APOSOR913219-1GEA 60.9 EUR
APOSOR913219-1G
5-Ethoxy-1H-indole-3-carbaldehyde 95%
5-Ethoxy-1H-indole-3-carbaldehyde
Formel: C11H11NO2
Molecular Weight: 189,2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07364715
CAS-Nummer: 169789-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR