5-Chloro-1H-indole-3-carbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-3-cyanoindole

OR913260-1G
APOSOR913260-1GEA 58 EUR
APOSOR913260-1G
5-Chloro-1H-indole-3-carbonitrile 95%
5-Chloro-1H-indole-3-carbonitrile
Formel: C₉H₅ClN₂
Molecular Weight: 176,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776618
CAS-Nummer: 194490-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR