5-Methyl-1H-indazole-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR913275-1G
OR913275-5G
APOSOR913275-1GEA
140
EUR
APOSOR913275-1G
APOSOR913275-5G
5-Methyl-1H-indazole-3-carbaldehyde 95%
5-Methyl-1H-indazole-3-carbaldehyde
Formel:
C₉H₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781624 CAS-Nummer: 518987-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...