5-Acetyl-2,3-dihydro-1H-indol-2-one 95%
Lieferant: Apollo Scientific
Synonyme:
5-Acetyloxindole
OR913281-1G
OR913281-5G
APOSOR913281-1GEA
78.3
EUR
APOSOR913281-1G
APOSOR913281-5G
5-Acetyl-2,3-dihydro-1H-indol-2-one 95%
5-Acetyl-2,3-dihydro-1H-indol-2-one
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782089 CAS-Nummer: 64483-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...