(S)-tert-Butyl(2-oxopiperidin-3-yl)carbamate

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-tert-Butyl (2-oxopiperidin-3-yl)carbamate, (3S)-3-Aminopiperidin-2-one, 3-BOC protected

OR913300-1G OR913300-5G
APOSOR913300-1GEA 36.4 EUR
APOSOR913300-1G APOSOR913300-5G
(S)-tert-Butyl(2-oxopiperidin-3-yl)carbamate
(S)-tert-Butyl(2-oxopiperidin-3-yl)carbamate
Formel: C₁₀H₁₈N₂O₃
Molecular Weight: 214.2600 g/mol
Schmelzpunkt: 100…103 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08166304
CAS-Nummer: 92235-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR