4-(2-Methoxyethoxy)aniline 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR913383-10G
OR913383-1G
OR913383-5G
APOSOR913383-10GEA
453
EUR
APOSOR913383-10G
APOSOR913383-1G
APOSOR913383-5G
4-(2-Methoxyethoxy)aniline 98%
4-(2-Methoxyethoxy)aniline
Formel:
C₉H₁₃NO₂ Molecular Weight: 167,2050 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03004838 CAS-Nummer: 33311-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...