(R)-(+)-Methylsuccinic acid 95%

Lieferant: Apollo Scientific

Synonyme: (R)-(+)-Pyrotartaric Acid

OR913598-1G
APOSOR913598-1GEA 55.1 EUR
APOSOR913598-1G
(R)-(+)-Methylsuccinic acid 95%
(R)-(+)-Methylsuccinic acid
Formel: C₅H₈O₄
Molecular Weight: 132,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209602
CAS-Nummer: 3641-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR