(S)-(-)-Methylsuccinic acid 95%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Methylsuccinic acid

OR913784-1G OR913784-5G
APOSOR913784-1GEA 49.3 EUR
APOSOR913784-1G APOSOR913784-5G
(S)-(-)-Methylsuccinic acid 95%
(S)-(-)-Methylsuccinic acid
Formel: C₅H₈O₄
Molecular Weight: 132.115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00192321
CAS-Nummer: 2174-58-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR