3-Imidazol-1-ylmethylphenylamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[(1H-Imidazol-1-yl)methyl]aniline

OR9140-1G OR9140-250MG
APOSOR9140-1GEA 357 EUR
APOSOR9140-1G APOSOR9140-250MG
3-Imidazol-1-ylmethylphenylamine 97%
3-Imidazol-1-ylmethylphenylamine
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,22 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07801129
CAS-Nummer: 120107-85-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR