8-Amino-2-methylchinolin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methylquinolin-8-amine, 8-Aminoquinaldine

OR914209-100G OR914209-25G
APOSOR914209-100GEA 427 EUR
APOSOR914209-100G APOSOR914209-25G
8-Amino-2-methylchinolin 98%
8-Amino-2-methylquinoline
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,1998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023998
CAS-Nummer: 18978-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR