2-Ethylthiazol 97%
Lieferant: Apollo Scientific
OR914220-1G
OR914220-25G
OR914220-5G
APOSOR914220-1GEA
125
EUR
APOSOR914220-1G
APOSOR914220-25G
APOSOR914220-5G
2-Ethylthiazol 97%
2-Ethylthiazole
Formel:
C₅H₇NS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085193 CAS-Nummer: 15679-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...