Ethyl-3-bromoisoxazole-5-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-bromoisoxazole-5-carboxylate

OR914272-1G OR914272-5G
APOSOR914272-1GEA 85.7 EUR
APOSOR914272-1G APOSOR914272-5G
Ethyl-3-bromoisoxazole-5-carboxylate 98%
Ethyl-3-bromoisoxazole-5-carboxylate
Formel: C₆H₆BrNO₃
Molecular Weight: 220,0200 g/mol
Schmelzpunkt: 47…48 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04035587
CAS-Nummer: 105174-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR