6-Bromo-1H-indole-3-carboxylic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR914279-1G
OR914279-5G
OR914279-10G
APOSOR914279-1GEA
33.5
EUR
APOSOR914279-1G
APOSOR914279-5G
APOSOR914279-10G
6-Bromo-1H-indole-3-carboxylic acid 98%
6-Bromo-1H-indole-3-carboxylic acid
Formel:
C₉H₆BrNO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD05664008 CAS-Nummer: 101774-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...