1-Isopropoxy-4-nitrobenzol 96%

Lieferant: Apollo Scientific

Synonyme: Isopropyl 4-Nitrophenyl Ether

OR914346-25G
APOSOR914346-25GEA 127 EUR
APOSOR914346-25G
1-Isopropoxy-4-nitrobenzol 96%
1-Isopropoxy-4-nitrobenzene
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01327671
CAS-Nummer: 26455-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR