3-Methyl-4-nitropyridin 98%

Lieferant: Apollo Scientific

Synonyme: 4-Nitro-3-picolin

OR914353-25G OR914353-5G
APOSOR914353-25GEA 672 EUR
APOSOR914353-25G APOSOR914353-5G
3-Methyl-4-nitropyridin 98%
3-Methyl-4-nitropyridin
Formel: C₆H₆N₂O₂
Molecular Weight: 138,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095084
CAS-Nummer: 1678-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR