5,6,7,8-Tetrahydro-1-naphthalinol 98%
Lieferant: Apollo Scientific
OR914367-100G
OR914367-25G
APOSOR914367-100GEA
278
EUR
APOSOR914367-100G
APOSOR914367-25G
5,6,7,8-Tetrahydro-1-naphthalinol 98%
5,6,7,8-Tetrahydro-1-naphthol
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001734 CAS-Nummer: 529-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...