2-Bromodiethylacetic acid 96%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-2-ethylbutanoic acid

OR914403-25G OR914403-5G
APOSOR914403-25GEA 502 EUR
APOSOR914403-25G APOSOR914403-5G
2-Bromodiethylacetic acid 96%
2-Bromodiethylacetic acid
Formel: C₆H₁₁BrO₂
Molecular Weight: 195.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00046512
CAS-Nummer: 5456-23-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR