2-Chlor-5-methylanisol 96%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-2-methoxy-4-methylbenzene

OR914539-1G OR914539-25G OR914539-5G
APOSOR914539-1GEA 125 EUR
APOSOR914539-1G APOSOR914539-25G APOSOR914539-5G
2-Chlor-5-methylanisol 96%
2-Chlor-5-methylanisol
Formel: C₈H₉ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD01679710
CAS-Nummer: 73909-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR